Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAPH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104059
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104059 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104059 Supplier Invitrogen™ Supplier No. PA5104059
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RAPH1.

This gene encodes the L protein of the mitochondrial glycine cleavage system. The L protein, also named dihydrolipoamide dehydrogenase, is also a component of the pyruvate dehydrogenase complex, the alpha-ketoglutarate dehydrogenase complex, and the branched-chain alpha-keto acide dehydrogenase complex. Mutations in this gene have been identified in patients with E3-deficient maple syrup urine disease and lipoamide dehydrogenase deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAPH1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RAPH1
Gene Accession No. Q70E73
Gene Alias 9430025M21Rik; ALS2CR18; ALS2CR9; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 18; amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 9; amyotrophic lateral sclerosis 2 chromosomal region candidate gene 18 protein; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 9 protein; C730009O10Rik; KIAA1681; Lamellipodin; LPD; PREL2; PREL-2; proline-rich EVH1 ligand 2; Protein RMO1; RalGDS/AF-6; RAPH1; Ras association (RalGDS/AF-6) and pleckstrin homology domains 1; ras-associated and pleckstrin homology domains-containing protein 1; RMO1
Gene Symbols RAPH1
Host Species Rabbit
Immunogen A synthesized peptide derived from human RAPH1(Accession Q70E73), corresponding to amino acid residues P931-I981.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65059
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.