Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAPH1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | RAPH1 |
---|---|
Applications | Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-0851
![]() |
Bethyl Laboratories, Inc
A305603AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
Lamellipodin, also called ras-associated and pleckstrin homology domains-containing protein 1 (RAPH1), is a 1302 amino acid member of the MRL family. The peripheral membrane protein mediates localized membrane signals and colocalizes at the tips of filopodia and lamellipodia with ENAH/VASP. Aside from the main isoform, nine additional isoforms have been identified for Lamellipodin (RMO1, RMO1a, RMO1b, RMO1c, RMO1ab, RMO1ac, RMO1bc, RMO1abc and RMO1-RAPH1). RMO1-RAPH1 is expressed in a wide variety of tissues, most highly in brain, heart, ovary and developing embryo. RMO1 is also widely expressed, with highest amounts in liver. Lamellipodin is down regulated in breast and ovarian cancers and shows reduced expression in metastatic osteosarcomas in comparison to primary osteosarcoma tumors.Specifications
RAPH1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Ras-associated and pleckstrin homology domains-containing protein 1, RAPH1, Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 18 protein, Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 9 protein, Lamellipodin, Proline-rich EVH1 ligand 2, PREL-2, Protein RMO1, RAPH1, ALS2CR18, ALS2CR9, KIAA1681, LPD, PREL2, RMO1 | |
RAPH1 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
Q70E73 | |
65059 | |
Between 1200 and 1250 | |
Primary | |
4° C | |
RAPH1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title