Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Germaine™ Laboratories Rapid Check™ Respiratory Syncytial Virus RSV Antigen Test

Catalog No. 23111441
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-111-441 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-111-441 Supplier Germaine™ Laboratories Supplier No. GCRSV502BUS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapid, qualitative detection of respiratory syncytial virus fusion protein in nasopharyngeal swabs/nasal aspirate specimens in children less than 6 years and adults over 60. Intended as an aid in the rapid laboratory diagnosis of acute respiratory syncytial virus infection in patients with such symptoms.

  • Viable air samplers are networked devices that can be remotely controlled and monitored
  • Strict regulation of air flow and will alarm the operator if the air flow deviates beyond the acceptable range
  • System also monitors and indicates a variety of operational parameters including sample's volume and elapsed time
  • Lightweight polymer
  • Surfaces compatible with chemical disinfection
  • Display graphics keep user informed in real time of air flow and remaining sample time
  • 8-hour run time
  • Touchscreen works with gloved hands

Specifications

Control Sets Positive and Negative Controls Included.
CPT Code 87807
CE Marker No
Clia Complexity Waived
Sample Type Nasopharyngeal Swabs/Nasal Aspirates
Sensitivity Premier Test Performance: Positive Percent Agreement: 93. 9%, Negative Percent Agreement: 97.7%
Type RSV Rapid Test
For Use With (Application) For Children Less than 6 Years Old and Adults Over 60 Years
Includes 20 Individually Packaged Test Devices, Nasopharyngeal Swabs with Each Kit, 1 R1 (Extraction Reagent Solution) 2 mL, 20 Extraction Tubes, 40 Disposable Pipettes, 1 PC (Positive Control Reagent) 1.0 mL, 1 NC (Negative Control Reagent) 1.0 mL, and 1 Instructions for Use.
Quantity 1 Kit
Test Time 15 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.