Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siemens Healthineers Rapid QC Plus Blood Gas Quality Control
Click to view available options
Level:
1
2
3
Description
- 30 ampules per level
- 2.5mL of organic salt buffered bicarbonate solution per ampule
- Yields expected values for pH/blood gas, Na+, K+, Cl- and Ca+ + (ionized and total calcium)

Specifications
Specifications
Type | Blood Gas Quality Control |
Intended Use | Used for yielding expected values for pH/Blood gas, Na+, K+, Cl-, Ca++ |
Level | 3 |
Quantity | 30 x 2.5 mL |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction