Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siemens Healthineers Rapid QC Plus Blood Gas Quality Control
Click to view available options
Level:
1
2
3
Description
- 30 ampules per level
- 2.5mL of organic salt buffered bicarbonate solution per ampule
- Yields expected values for pH/blood gas, Na+, K+, Cl- and Ca+ + (ionized and total calcium)

Specifications
Specifications
Type | Blood Gas Quality Control |
Intended Use | Used for yielding expected values for pH/Blood gas, Na+, K+, Cl-, Ca++ |
Level | 2 |
Quantity | 30 x 2.5 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction