Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Siemens Healthineers RAPIDLab™ 1200 Series Ready Sensor™ Biosensors/Electrodes
Click to view available options
Sensor Types:
Calcium Biosensor
Chloride Biosensor
Lactate Biosensor
Potassium Biosensor
Sodium Biosensor
pCO2 Biosensor
pH Biosensor
pO2 Biosensor
Specifications
Specifications
Sensor Types | Calcium Biosensor |
For Use With (Equipment) | RAPIDLab™ 1200 Series Instruments |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction