Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ RapidPure™ RNA Plant Kit

Catalog No. mp112722050 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP112722050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP112722050 Supplier MP Biomedicals™ Supplier No. 112722050

Isolate and purify high-quality total RNA from plant cells, plant tissues and filamentous fungi. MP Biomedicals RapidPure™ RNA Plant Kit isolates up to 80μg total RNA from up to 100mg or 1x107 cells of material.

Designed to isolate and purify RNA from plant cells, plant tissues and filamentous fungi in a spin filter format. Special buffer conditions guarantee an efficient lysis of the starting material and a simultaneous inactivation of endogenous RNases. Applications include Northern Blotting, cDNA transcription, real-time RT-PCR, DDRT-PCR, array technologies, TaqMan™ analysis and many others.

TRUSTED_SUSTAINABILITY

Specifications

Cell Lines Tested Allium cepa, Arabidopsis thaliana, Ascomyceten, Brassica oleracea, Casuarina (Australian tree), Fusarium avenaceum, Gossypium spec., Lycopersicon esculentum, Malus sylvestris, Mangifera indica, Nicotiana tabacum, Oryza sativa, Picea abies, Rosa hybrida, Solanaceae, Vitis vinifera, Zea mays, Actinidia deliciosa, Beta vulgaris, Brassica napus, Chlorophyceae, Fungi, Hordeum vulgare, Solanum tuberosum
Content And Storage Store at room temperature.
Format Spin Column
For Use With (Application) Isolation and purification of high quality total RNA from plant cells, plant tissues and filamentous fungi
Includes Lysis Solution PS, Lysis Solution PH, RNA Wash Buffer 1, RNA Wash Buffer 2, RNA Elution Buffer, Prefilter, RNA Spin Filter Set, 2.0 mL Collection Tubes, 1.5 mL Elution Tubes
Quantity 50 Preps
Sample Size Less than or equal to 100 mg
Synonym Plant RNA Isolation Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.