Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ RapidScript™ RT Kit

Catalog No. MP116187600 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
No. of Reactions: 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MP116187600 No. of Reactions: 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116187600 Supplier MP Biomedicals™ Supplier No. 116187600
Add to Cart
Add to Cart

Use the RapidScript™ RT Kit to cDNA synthesize from 15 kb RNA templates in 10 to 15 min.

This specially engineered enzyme offers longer full-length cDNA transcripts, up to 12 kb, with excellent performance at high temperatures (50 to 60°C) to facilitate removal of secondary structures.

  • RNase H activity removed to achieve thermal stability
  • Ideal for use with difficult RNA templates
  • High yields and high-quality cDNA transcription
  • Primers and dNTPs not included
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Reverse Transcriptase, 100 μL; 5X RT Buffer, 400 μL. Store at -20°C
Form Liquid
Format Kit
PCR Method RT-PCR
Technique Reverse Transcription
For Use With (Equipment) Standard and Fast Cycling Instruments
Kit Type Reverse Transcription Kit
Label or Dye Unlabeled
No. of Tests 100 Test
Optimal Reaction Temperature 50°C to 60°C
Product Line RapidScript
Research Category PCR, qPCR, Gene Expression Analysis
Reverse Transcriptase M-MLV
Ribonuclease H Activity No
Sufficient For 100 Reactions
For Use With (Application) RT-PCR
Compatibility RT-PCR
Final Product Type cDNA
No. of Reactions 100 Reactions
Reaction Format Separate Components
Reagent Type Reverse Transcription
Sample Type RNA
Size (Final Product) Up to 12 kb
Starting Material RNA
Includes Reverse Transcriptase (100 μL), 5X RT Buffer (400 μL)
Show More Show Less

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.