Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Raptor Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520126
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520126 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520126 Supplier Invitrogen™ Supplier No. PA520126
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is L1210 cell lysate. PA5-20126 can be used with blocking peptide PEP-0244.

Regulatory associated protein of mTOR (RAPTOR) is a binding partner of target of rapamycin (TOR). It binds to 4EBP1 and p70alpha. The binding of raptor to mTOR is essential for the mTOR-catalyzed phosphorylation of 4EBP1 in vitro. It enhances the mTOR kinase activity toward p70alpha. It is also an essential scaffold for the mTOR-catalyzed phosphorylation of 4EBP1 and mediates TOR action in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Raptor
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene RPTOR
Gene Accession No. Q8K4Q0, Q8N122
Gene Alias 4932417H02Rik; KIAA1303; KOG1; Mip1; mKIAA1303; p150 target of rapamycin (TOR)-scaffold protein; p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats; Rap; RAPTOR; regulatory associated protein of MTOR complex 1; regulatory associated protein of MTOR, complex 1; regulatory-associated protein of mTOR; RGD1311784; RPTOR
Gene Symbols RPTOR
Host Species Rabbit
Immunogen A 15 amino acid peptide from the central region of human Raptor.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57521, 74370
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.