Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Raptor Recombinant Rabbit Monoclonal Antibody (1B10)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA549939
Description
Raptor Recombinant Monoclonal Antibody for IHC (P), ELISA
Regulatory associated protein of mTOR (RAPTOR) is a binding partner of target of rapamycin (TOR). It binds to 4EBP1 and p70alpha. The binding of raptor to mTOR is essential for the mTOR-catalyzed phosphorylation of 4EBP1 in vitro. It enhances the mTOR kinase activity toward p70alpha. It is also an essential scaffold for the mTOR-catalyzed phosphorylation of 4EBP1 and mediates TOR action in vivo.
Specifications
Raptor | |
Recombinant Monoclonal | |
1.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8N122 | |
RPTOR | |
A synthesized peptide derived from human Raptor. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
1B10 | |
Unconjugated | |
RPTOR | |
4932417H02Rik; KIAA1303; KOG1; Mip1; mKIAA1303; p150 target of rapamycin (TOR)-scaffold protein; p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats; Rap; RAPTOR; regulatory associated protein of MTOR complex 1; regulatory associated protein of MTOR, complex 1; regulatory-associated protein of mTOR; RGD1311784; RPTOR | |
Rabbit | |
Affinity chromatography | |
RUO | |
57521 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction