Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAR alpha/NR1B1 Antibody (OTI2C3), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273802AF405
Description
RAR alpha/NR1B1 Monoclonal specifically detects RAR alpha/NR1B1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
RAR alpha/NR1B1 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RARA | |
Recombinant protein fragment corresponding to amino acids 52-299 of human RAR alpha/NR1B1 (NP_000955) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2C3 | |
Western Blot, Immunohistochemistry | |
alpha polypeptide, RAR-alpha, retinoic acid receptor, alpha | |
Mouse | |
Affinity Purified | |
Cancer, Phospho Specific, Vision | |
5914.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction