Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAR beta/NR1B2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220129
Description
RAR beta/NR1B2 Polyclonal specifically detects RAR beta/NR1B2 in Human samples. It is validated for Western Blot.Specifications
RAR beta/NR1B2 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
beta polypeptide, HBV-activated protein, retinoic acid receptor, beta | |
Rabbit | |
50 kDa | |
0.1 mL | |
Cancer, Neuroscience, Sensory Systems, Transcription Factors and Regulators, Vision | |
5915 | |
Human | |
IgG |
Western Blot | |
0.97mg/mL | |
Western Blot 1:500-1:3000 | |
P10826 | |
RARB | |
Recombinant protein encompassing a sequence within the center region of human Retinoic Acid Receptor beta. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Species reactivity based on sequence identity: Mouse (98%), Chicken (96%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction