Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAR beta/NR1B2 Antibody (SR1998), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32165450UL
Description
RAR beta/NR1B2 Monoclonal antibody specifically detects RAR beta/NR1B2 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
RAR beta/NR1B2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
SR1998 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
beta polypeptide, HBV-activated protein, retinoic acid receptor, beta | |
A synthesized peptide derived from human RAR beta/NR1B2 (Uniprot #: P10826) | |
50 μg | |
Cancer, Neuroscience, Sensory Systems, Transcription Factors and Regulators, Vision | |
5915 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction