Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAR beta/NR1B2 Rabbit anti-Human, Mouse, Clone: 6D5V4, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
| Antigen | RAR beta/NR1B2 |
|---|---|
| Clone | 6D5V4 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
RAR beta/NR1B2 Monoclonal antibody specifically detects RAR beta/NR1B2 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| RAR beta/NR1B2 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Neuroscience, Sensory Systems, Transcription Factors and Regulators, Vision | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 5915 | |
| IgG | |
| Affinity purified |
| 6D5V4 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| beta polypeptide, HBV-activated protein, retinoic acid receptor, beta | |
| A synthetic peptide corresponding to a sequence within amino acids 350-455 of human RAR beta/NR1B2 (P10826). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title