Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAR Beta Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980323
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0323 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0323 Supplier Bioss Supplier No. BS0516R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Retinoic Acid (RA; active metabolite of vitamin A) plays a prominent role in regulating the transition of proliferating precursor cells (such as carcinoma cells and neuronal precursors) to postmitotic differentiated cells. The Retinoid X Receptors (RXRs) family (RXR alpha, beta, and gamma), preferentially bind 9-cis -RA and regulate gene transcription by forming heterodimers with a second family of RA receptors. RAs have been suggested to potentially play a therapeutic role in cervical cancer. RAs are known to play key roles in neuronal development and an increasing body of evidence indicates that retinoid signaling may regulate synaptic plasticity and associated learning and memory behaviors.

Specifications

Antigen RAR Beta
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RARB
Gene Accession No. P10826
Gene Alias A830025K23; HAP; HBV-activated protein; hepatitis B virus activated protein; MCOPS12; NR1B2; Nuclear receptor subfamily 1 group B member 2; RAR beta; RARB; RAR-b; RAR-beta; RARbeta1; RAR-Beta2; RAR-Beta4; RAR-epsilon; Retinoic acid receptor beta; retinoic acid receptor, beta; retinoic acid receptor, beta polypeptide; RRB2
Gene Symbols RARB
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse RAR Beta .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5915
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.