Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAR gamma-1,2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121101 Supplier Invitrogen™ Supplier No. PA5121101
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse lung, Rat kidney. The target is usually found in the following locations: Nucleus. Immunogen sequence: KKEVKEEGSP DSYELSPQLE ELITKVSKAH QE.

Retinoic acid, a metabolite of vitamin A, is necessary for normal organogenesis but acts as a teratogen at high levels during embryonic and fetal development. Retinoic acid functions through its interaction with the nuclear protein, retinoic acid receptor (RAR). RAR belongs to the steroid and thyroid hormone superfamily of nuclear receptor proteins which exert their effects by binding to specific DNA response elements, thus regulating gene expression in target cells. RAR exists as three major subtypes: alpha, beta and gamma. Two main isoforms of RAR gamma exist: RAR gamma-1 has been found to be predominantly expressed in skin, while RAR gamma-2 has been found early in embryogenesis and in embryonal carcinoma cells. RAR gamma is also known as RARD. The corresponding gene for the retinoic acid receptor gamma 1 is NR1B3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAR gamma-1,2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.82 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RARG
Gene Accession No. P13631, P18911
Gene Symbols RARG
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 168-199 of human RARG (NP_0009571).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19411, 5916, 685072
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.