Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAR gamma-1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA3813
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA3813 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA3813 Supplier Invitrogen™ Supplier No. PA3813
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA3-813 detects recombinant human retinoic acid receptor (RAR) gamma-1. The immunizing sequence is 100% conserved in mouse. This antibody detects RAR gamma-1, but not RAR gamma-2, or either of the alpha or beta forms of RAR. PA3-813 has been successfully used in Western blot, immunoprecipitation and gel shift procedures. By Western blot, this antibody detects an ~50 kDa protein representing RAR gamma-1 from transiently transfected 293T cells. PA3-813 has been shown to efficiently super shift the human RAR gamma-1/retinoid X receptor (RXR)/RAR response element complex in gel super shift experiments using a number of different response elements. PA3-813 immunizing peptide corresponds to the N-terminus of RAR gamma-1. This peptide (Cat. # PEP-228) is available for use in neutralization and control experiments.

Retinoic acid, a metabolite of vitamin A, is necessary for normal organogenesis but acts as a teratogen at high levels during embryonic and fetal development. Retinoic acid functions through its interaction with the nuclear protein, retinoic acid receptor (RAR). RAR belongs to the steroid and thyroid hormone superfamily of nuclear receptor proteins which exert their effects by binding to specific DNA response elements, thus regulating gene expression in target cells. RAR exists as three major subtypes: alpha, beta and gamma. Two main isoforms of RAR gamma exist: RAR gamma-1 has been found to be predominantly expressed in skin, while RAR gamma-2 has been found early in embryogenesis and in embryonal carcinoma cells. RAR gamma is also known as RARD. The corresponding gene for the retinoic acid receptor gamma 1 is NR1B3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAR gamma-1
Applications Gel Shift, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum, PBS with 0.05% sodium azide
Gene Rarg
Gene Accession No. P13631, P18911
Gene Alias Nr1b3; Nuclear receptor subfamily 1 group B member 3; RAR gamma 2; RAR Gamma-1; RARC; RARD; Rarg; RAR-gamma; RARgamma2; retinoic acid; retinoic acid nuclear receptor gamma variant 1; retinoic acid nuclear receptor gamma variant 2; retinoic acid receptor; retinoic acid receptor gamma; retinoic acid receptor gamma 4; retinoic acid receptor, gamma
Gene Symbols Rarg
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the residues E(40) M L S P S F R G L G Q P D L P K E(57) of the N-terminal end of RAR gamma-1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19411, 5916, 685072
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.