Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RARA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587426
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587426 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587426 Supplier Invitrogen™ Supplier No. PA587426
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MYESVEVGGP TPNPFLVVDF YNQNRACLLP EKGLPAPGPY STPLRTPLWN GSNHSIETQS SSSEEIVPSP PSPPPLPRIY KPCFVCQDKS SGYHYGVSAC EGCKGFFRRS IQKNMVYTCH RDKNCIINKV TRNRCQYCRL QKCFEVGMSK ESVRNDRNKK KKEVPKPECS ESYTLTPEVG ELIEKVRKAH QETFPALCQL GKYTTNNSSE QRVSLDIDLW DKFSELSTKC IIKTVEFAKQ LPGFTTLTIA DQITLLKAAC LDILILRICT RYTPEQDTMT FSDGLTLNRT QMHNAGFGPL TDLVFAFANQ LLPLEMDDAE TGLLSAICLI CGDRQDLEQP DRVDMLQEPL LEALKVYVRK RRPSRPHMFP KMLMKITDLR SISAKGAERV ITLKMEIPGS MPPLIQEMLE NSEGLDTLSG QPGGGGRDGG GLAPPPGSCS PSLSPSSNRS SPATHSP; Positive Samples: MCF7, HeLa, HepG2, NIH/3T3, C6, Rat heart, Rat testis; Cellular Location: Cytoplasm, Nucleus.

RARA is a receptor for retinoic acid. Retinoic acid receptors bind as heterodimers to their target response elements in response to their ligands, all-trans or 9-cis retinoic acid, and regulate gene expression in various biological processes. The RXR/RAR heterodimers bind to the retinoic acid response elements (RARE) composed of tandem 5'-AGGTCA-3' sites known as DR1-DR5. In the absence of ligand, the RXR-RAR heterodimers associate with a multiprotein complex containing transcription corepressors that induce histone acetylation, chromatin condensation and transcriptional suppression. On ligand binding, the corepressors dissociate from the receptors and associate with the coactivators leading to transcriptional activation. RARA plays an essential role in the regulation of retinoic acid-induced germ cell development during spermatogenesis. Has a role in the survival of early spermatocytes at the beginning prophase of meiosis. In Sertoli cells, may promote the survival and development of early meiotic prophase spermatocytes. In concert with RARG, required for skeletal growth, matrix homeostasis and growth plate function. Regulates expression of target genes in a ligand-dependent manner by recruiting chromatin complexes containing KMT2E/MLL5. Mediates retinoic acid-induced granulopoiesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RARA
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RARA
Gene Accession No. P10276, P11416
Gene Alias Nr1b1; nuclear receptor subfamily 1 group B member 1; nucleophosmin-retinoic acid receptor alpha fusion protein NPM-RAR long form; RAR; RAR Alpha; RAR alpha 1; RARA; RAR-alpha; RARalpha1; retinoic acid; retinoic acid nuclear receptor alpha variant 1; retinoic acid nuclear receptor alpha variant 2; retinoic acid receptor; retinoic acid receptor alpha; retinoic acid receptor, alpha; retinoic acid receptor, alpha polypeptide; RRA
Gene Symbols RARA
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-457 of human RARa (NP_0010199801).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19401, 24705, 5914
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.