Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RARRES1 Antibody (OTI1D2), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273803G
Description
RARRES1 Monoclonal specifically detects RARRES1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
RARRES1 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RARRES1 | |
Full length recombinant protein of human RARRES1 (NP_996846) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG2b |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1D2 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
RAR-responsive protein TIG1, retinoic acid receptor responder (tazarotene induced) 1, retinoic acid receptor responder protein 1, TIG1Tazarotene-induced gene 1 protein | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
5918.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction