Learn More
Invitrogen™ RARRES1 Monoclonal Antibody (OTI1B8)

Description
RARRES1 Monoclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antigen | RARRES1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI1B8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RARRES1 |
| Gene Accession No. | P49788 |
| Gene Alias | 5430417P09Rik; AI662122; latexin-like; LXNL; PEIG1; PERG-1; phorbol ester-induced gene 1 protein; Rarres1; RAR-responsive protein TIG1; retinoic acid receptor responder (tazarotene induced) 1; retinoic acid receptor responder 1; retinoic acid receptor responder protein 1; Tazarotene-induced gene 1 protein; tazarotene-induced protein 1; TIG1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.