Learn More
RARRES1 Mouse anti-Human, Clone: OTI1H2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF506087

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene was identified as a retinoid acid (RA) receptor-responsive gene. It encodes a type 1 membrane protein. The expression of this gene is upregulated by tazarotene as well as by retinoic acid receptors. The expression of this gene is found to be downregulated in prostate cancer, which is caused by the methylation of its promoter and CpG island. Alternatively spliced transcript variant encoding distinct isoforms have been observed.Specifications
RARRES1 | |
Monoclonal | |
Unconjugated | |
RARRES1 | |
LXNL, PERG-1, TIG1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
5918 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1H2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P49788 | |
RARRES1 | |
Full length human recombit protein of human RARRES1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.