Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RARRES1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547645
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547645 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547645 Supplier Invitrogen™ Supplier No. PA547645
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human RARRES1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

This gene was identified as a retinoid acid (RA) receptor-responsive gene. It encodes a type 1 membrane protein. The expression of this gene is upregulated by tazarotene as well as by retinoic acid receptors. The expression of this gene is found to be downregulated in prostate cancer, which is caused by the methylation of its promoter and CpG island. Alternatively spliced transcript variant encoding distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RARRES1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene RARRES1
Gene Accession No. 0
Gene Alias 5430417P09Rik; AI662122; latexin-like; LXNL; PEIG1; PERG-1; phorbol ester-induced gene 1 protein; Rarres1; RAR-responsive protein TIG1; retinoic acid receptor responder (tazarotene induced) 1; retinoic acid receptor responder 1; retinoic acid receptor responder protein 1; Tazarotene-induced gene 1 protein; tazarotene-induced protein 1; TIG1
Gene Symbols RARRES1
Host Species Goat
Immunogen E. coli-derived recombinant mouse RARRES1 Pro78-Phe283.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109222
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.