Learn More
Invitrogen™ RARRES3 Monoclonal Antibody (659209)

Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Specifications
Specifications
| Antigen | RARRES3 |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 659209 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | PLAAT4 |
| Gene Accession No. | Q9UL19 |
| Gene Alias | HRAS-like suppressor 4; HRASLS4; HRSL4; Phospholipase A and acyltransferase 4; PLA1/2-3; PLAAT4; RARRES3; RAR-responsive protein TIG3; retinoic acid receptor responder (tazarotene induced) 3; retinoic acid receptor responder 3; retinoic acid receptor responder protein 3; retinoic acid-inducible gene 1; retinoid-inducible gene 1 protein; RIG1; Tazarotene-induced gene 3 protein; TIG3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.