Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RARRES3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597156
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597156 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597156 Supplier Invitrogen™ Supplier No. PA597156
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MASPHQEPKP GDLIEIFRLG YEHWALYIGD GYVIHLAPPS EYPGAGSSSV FSVLSNSAEV KRERLEDVVG GCCYRVNNSL DHEYQPRPVE VIISSAKEMV GQKMKYSIVS RNCEHFVTQL RYGKSRCKQV EKAKV; Positive Samples: 293T; Cellular Location: Membrane, Single-pass membrane protein.

Retinoids exert biologic effects such as potent growth inhibitory and cell differentiation activities and are used in the treatment of hyperproliferative dermatological diseases. These effects are mediated by specific nuclear receptor proteins that are members of the steroid and thyroid hormone receptor superfamily of transcriptional regulators. RARRES1, RARRES2, and RARRES3 are genes whose expression is upregulated by the synthetic retinoid tazarotene. RARRES3 is thought act as a tumor suppressor or growth regulator.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RARRES3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PLAAT4
Gene Accession No. Q9UL19
Gene Alias HRAS-like suppressor 4; HRASLS4; HRSL4; Phospholipase A and acyltransferase 4; PLA1/2-3; PLAAT4; RARRES3; RAR-responsive protein TIG3; retinoic acid receptor responder (tazarotene induced) 3; retinoic acid receptor responder 3; retinoic acid receptor responder protein 3; retinoic acid-inducible gene 1; retinoid-inducible gene 1 protein; RIG1; Tazarotene-induced gene 3 protein; TIG3
Gene Symbols PLAAT4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-135 of human RARRES3 (NP_0045762).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5920
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.