Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RARS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110272
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110272 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110272 Supplier Invitrogen™ Supplier No. PA5110272
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LNLPPENLIT SISAVPISQK EEVADFQLSV DSLLEKDNDH SRPDIQVQAK RLAEKLRCDT VVSEISTGQR TVNFKINREL LTKTVLQQVI EDGSKYGLKS ELFSGLPQKK IVVEFSSPNV AKKFHVGHLR STIIGNFIAN LKEALGHQVI RINYLGDWGM QFGLLGTGFQ LFGYEEKLQS NPLQHLFEVY VQVNKEAADD KSVAKAAQEF FQRLELGDVQ ALSLWQKFRD LSIEEYIRVY KRLGVYFDEY SGESFYREKS QEVLKLLESK GLLLKTIKGT AVVDLSGNGD PSSICTVMRS DGTSLYATRD LAAAIDRMDK YNFD.

The protein encoded by this gene is an arginyl-tRNA synthetase that is found in the mitochondrial matrix. Defects in this gene are a cause of pontocerebellar hypoplasia type 6 (PCH6).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RARS2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RARS2
Gene Accession No. Q3U186, Q5T160
Gene Alias 1500002I10Rik; arginine--tRNA ligase; Arginyl-tRNA synthetase; arginyl-tRNA synthetase 2, mitochondrial; arginyl-tRNA synthetase 2, mitochondrial (putative); ArgRS; DALRD2; dJ382I10.6; PCH6; PRO1992; probable arginine--tRNA ligase, mitochondrial; probable arginyl-tRNA synthetase, mitochondrial; Rars2; RARSL
Gene Symbols RARS2
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109093, 57038
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.