Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RARS2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572536
Description
RARS2 is an arginyl-tRNA synthetase that is found in the mitochondrial matrix.
Specifications
RARS2 | |
Polyclonal | |
Unconjugated | |
RARS2 | |
Probable arginine--tRNA ligase; mitochondrial; Arginyl-tRNA synthetase; ArgRS; RARS2; RARSL | |
Rabbit | |
selective precipitation | |
RUO | |
57038 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
5.77 mg/mL | |
PBS with 0.09% sodium azide | |
Q5T160 | |
RARS2 | |
KLH conjugated synthetic peptide between 369-399 amino acids from the C-terminal region of human RARS2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction