Learn More
Invitrogen™ Ras (G12V Mutant) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578086
Description
Positive Control: NRAS(G12V)-transfected 293T Predicted Reactivity: Mouse (100%), Rat (100%), Zebrafish (100%), Xenopus laevis (100%), Dog (100%), Pig (100%), Chicken (100%), Bovine (100%), Guinea pig (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Ras is an N-ras oncogene encoding a membrane protein that shuttles between the Golgi apparatus and the plasma membrane. The process of shuttling is regulated through palmitoylation and depalmitoylation by the ZDHHC9-GOLGA7 complex. The encoded protein, which has intrinsic GTPase activity, is activated by a guanine nucleotide-exchange factor and inactivated by a GTPase activating protein. Mutations in Ras gene have been associated with somatic rectal cancer, follicular thyroid cancer, autoimmune lymphoproliferative syndrome, Noonan syndrome, and juvenile myelomonocytic leukemia.
Specifications
Ras (G12V Mutant) | |
Polyclonal | |
Unconjugated | |
NRAS | |
ALPS4; CMNS; GTPase NRas; HRAS1; NCMS; neuroblastoma RAS viral (v-ras) oncogene homolog; neuroblastoma RAS viral oncogene homolog; NRAS; N-ras; N-ras protein part 4; NRAS1; NS6; Transforming protein N-Ras; v-ras neuroblastoma RAS viral oncogene homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4893 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1.02 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
P01111 | |
NRAS | |
Carrier-protein conjugated synthetic peptide surrounding mutant G12V of human K-Ras. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.