Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ras-GAP Antibody (OTI1H2) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273804
Description
Ras-GAP Monoclonal specifically detects Ras-GAP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
RAS p21 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
CMAVM, DKFZp434N071, GAPp120GAPCM-AVM, GTPase-activating protein, p120RASGAP, PKWS, ras GTPase-activating protein 1, Ras p21 protein activator, RAS p21 protein activator (GTPase activating protein) 1, RASA, RasGAP, triphosphatase-activating protein | |
Mouse | |
116.2 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
OTI1H2 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
RASA1 | |
Full length human recombinant rotein of human RASA1(NP_002881) produced in HEK293T cell. | |
Affinity Purified | |
Cancer, Core ESC Like Genes, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers | |
5921 | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction