Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ras Monoclonal Antibody (4E5), Abbomax

Catalog No. 502204138
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4138 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4138 Supplier Abbomax Inc Supplier No. 603090
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Ras full length protein. Cellular location: Cell membrane, Golgi apparatus membrane.

Ras Monoclonal antibody specifically detects Ras in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Ras
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 400000
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Nras
Gene Accession No. P01111, P01112, P01116
Gene Alias AI929937; ALPS4; AV095280; C-BAS/HAS; cellular c-Ki-ras2; cellular c-Ki-ras2 proto-oncogene; CFC2; c-Ha-ras; c-Ha-ras p21 protein; c-Ha-ras transgene; C-HA-RAS1; c-has/bas p21 protein; C-H-RAS; c-Ki-ras; c-Kirsten-ras protein; c-Kirsten-ras proto-oncogene; c-K-ras; c-K-ras2 protein; CMNS; c-rasHa; c-ras-Ki-2 activated oncogene; CTLO; etID18709.8; fa04e08; fc14b12; fc23g10; fj89d12; fj99a10; GTP- and GDP-binding peptide B; GTPase HRas; GTPase HRas, N-terminally processed; GTPase Hras1; GTPase KRas; GTPase KRas, N-terminally processed; GTPase NRas; HAMSV; Ha-ras; Ha-Ras1 proto-oncoprotein; Harvey ras1 protein; Harvey rat sarcoma viral (v-Ha-ras) oncogene homolog; Harvey rat sarcoma viral oncogene homolog; Harvey rat sarcoma viral oncoprotein; Harvey rat sarcoma virus oncogene; Harvey rat sarcoma virus oncogene 1; Harvey-ras; hras; H-ras; H-ras 1 protein; HRas proto-oncogene, GTPase; HRas proto-oncogene, GTPase a; HRAS1; H-Ras-1; Hras-1; hrasa; H-RASIDX; hypothetical protein LOC445289; hypothetical protein LOC550286; K RAS; Ki-ras; Kirsten rat sarcoma oncogene 2, expressed; Kirsten rat sarcoma viral oncogene; Kirsten rat sarcoma viral oncogene homolog; Kirsten rat sarcoma viral oncogene homologue 2 (active); Kras; K-ras; K-Ras 2; K-ras p21 protein; KRAS proto-oncogene, GTPase; KRAS1; KRAS2; Kras-2; K-RAS2A; K-RAS2B; K-RAS4A; K-RAS4B; NCMS; Neuroblastoma RAS; neuroblastoma ras oncogene; neuroblastoma RAS viral (v-ras) oncogene homolog; neuroblastoma RAS viral oncogene homolog; NRAS; N-ras; N-ras oncogene p21; N-ras protein part 4; NRAS proto-oncogene, GTPase; NRAS1; NS; NS3; NS6; oncogene KRAS2; p19 H-RasIDX protein; p21; p21 N-ras oncogene; p21 protein; p21B; p21ras; PR310 c-K-ras oncogene; RALD; ras; Ras family small GTP binding protein H-Ras; RASH1; RASK; RASK2; ras-like protein; RASN; sb:eu826; transformation gene: oncogene HAMSV; transforming protein N-Ras; Transforming protein p21; v-Ha-ras Harvey rat sarcoma viral oncogene homolog; v-Ha-ras Harvey rat sarcoma viral oncogene homolog a; v-Ki-ras2 Kirsten rat sarcoma 2 viral oncogene homolog; v-Ki-ras2 Kirsten rat sarcoma viral oncogene homolog; v-ras neuroblastoma RAS viral oncogene homolog; wu:fa04e08; wu:fc14b12; wu:fc23g10; wu:fj89d12; wu:fj99a10; xRAS1; xRAS2; zgc:100761; zgc:110250; zgc:85725; zras-b1
Host Species Mouse
Immunogen A full length of human Ras recombinant protein
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3265, 3845, 4893
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.