Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Ras, Mouse, Alexa Fluor 555, Clone: RAS10 ,

Catalog No. 05516AF555 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 555
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
05516AF555 100 μL Alexa Fluor 555
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05516AF555 Supplier MilliporeSigma™ Supplier No. 05516AF555
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ras proteins are small GTPases controlling cellular pathways that signal for growth, proliferation, and differentiation. Mammalian genomes harbor three Ras genes (HRAS, NRAS, and KRAS) that encode four GTPases, two of which (K-Ras4A and K-Ras4B) are transcribed from the KRAS gene via two alternative fourth exons. Ras proteins share nearly identical sequence homology in sequence encoded by exons 1-3 (a.a. 1-165), while diverge in their C-terminal 24-a.a. hypervariable region (HVR) encoded by exon 4. The first 165 amino acids form the catalytic G domain that mediates GDP/GTP binding and association with effectors, exchange factors, and GTPase-activating proteins (GAPs), while the C-terminal HVR is needed for Ras membrane localization, a process necessary for Ras function. In addition to the removal of initiator methionine (Met1), Ras proteins are posttranslationally modified by farnesylation of Cys186, followed by proteolytical cleavage their C-terminal end three amino acids (a.a. 187-189) and carboxyl methylation of the newly formed C-terminal prenylcysteine. H-Ras, N-Ras, and K-Ras4A are further palmitoylated on cysteine residue(s) near the C-terminus (Cys181/Cys184 in H-Ras, Cys181 in N- and K-Ras), while K-Ras4B lacks a palmitoylation site. Palmitoylation is required for targeting H- and N-Ras to the plasma membrane (PM), while K-Ras4A and K-Ras4B contain a series of lysines that constitute a polybasic region (PBR) capable of interacting with the inner leaflet of the PM through an electrostatic interaction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ras
Applications Immunocytochemistry
Classification Monoclonal
Clone RAS10
Concentration Please refer to lot specific datasheet.
Conjugate Alexa Fluor 555
Formulation Purified mouse IgG2a Alexa Fluor™ 555 conjugate in buffer containing PBS with 1.5% BSA and 0.05% Sodium Azide.
Gene HRAS, HRAS1, KRAS, CFC2, NS3, KRAS2, RASK2, NRAS, NS6, CMNS, NCMS
Gene Accession No. P01112
Host Species Mouse
Immunogen Recombinant human H-Ras p21 (Hamer, P.J., et al. (1990). Hybridoma.9(6):573-587).
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NM_001130442;NM_005343
Test Specificity Ras 10 reacts with normal p21, activated p21, and p21 from each of the Harvey, Kirsten and N-ras families. Ras 10 reacts with recombinant p21 as well as p21 derived from human and rodent cells (Hamer, P.J., et al. (1990). Hybridoma.9(6):573-587).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.