Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAS p21 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP259127
Description
Ras-GAP Polyclonal specifically detects Ras-GAP in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation (Negative).Specifications
RAS p21 | |
Polyclonal | |
Western Blot 1:1000, Immunoprecipitation (Negative) | |
CMAVM, DKFZp434N071, GAPp120GAPCM-AVM, GTPase-activating protein, p120RASGAP, PKWS, ras GTPase-activating protein 1, Ras p21 protein activator, RAS p21 protein activator (GTPase activating protein) 1, RASA, RasGAP, triphosphatase-activating protein | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide | |
RASA1 | |
Maps to a region between 625 and 675 | |
100 μL | |
Cancer, Core ESC Like Genes, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers | |
5921 | |
Store at 4C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction