Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ras (Q61R Mutant) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578093
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578093 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578093 Supplier Invitrogen™ Supplier No. PA578093
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Human Ras (Q61R Mutant specific)-transfected 293T whole cell extracts Predicted Reactivity: Mouse (100%), Rat (100%), Zebrafish (100%), Xenopus laevis (100%), Dog (100%), Pig (100%), Chicken (100%), Bovine (100%), Guinea pig (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Ras is an N-ras oncogene encoding a membrane protein that shuttles between the Golgi apparatus and the plasma membrane. The process of shuttling is regulated through palmitoylation and depalmitoylation by the ZDHHC9-GOLGA7 complex. The encoded protein, which has intrinsic GTPase activity, is activated by a guanine nucleotide-exchange factor and inactivated by a GTPase activating protein. Mutations in Ras gene have been associated with somatic rectal cancer, follicular thyroid cancer, autoimmune lymphoproliferative syndrome, Noonan syndrome, and juvenile myelomonocytic leukemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ras (Q61R Mutant)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene NRAS
Gene Accession No. P01111
Gene Alias ALPS4; CMNS; GTPase NRas; HRAS1; NCMS; neuroblastoma RAS viral (v-ras) oncogene homolog; neuroblastoma RAS viral oncogene homolog; NRAS; N-ras; N-ras protein part 4; NRAS1; NS6; Pan Ras; Transforming protein N-Ras; v-ras neuroblastoma RAS viral oncogene homolog
Gene Symbols NRAS
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide surrounding mutant Q61R of human K-Ras. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4893
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.