Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ras Rabbit anti-Human, Mouse, Rat, Clone: 3G8L8, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $515.50
Specifications
| Antigen | Ras |
|---|---|
| Clone | 3G8L8 |
| Applications | Western Blot, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
Ras Monoclonal antibody specifically detects Ras in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
| Ras | |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cellular Signaling, Epigenetics | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| IgG | |
| Affinity purified |
| 3G8L8 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| A synthesized peptide derived from human KRAS+HRAS+NRAS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title