Learn More
RASA1 Mouse anti-Human, Clone: OTI1H2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505899

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
GTPase Activating Protein (GAP) is capable of stimulating the hydrolysis of GTP by ras p21 proteins, though GAP has little effect on the oncogenic forms of ras. It is also known that several tyrosine kinases such as platelet derived growth factor receptor and epidermal growth factor receptor are involved in the tyrosine phosphorylation of GAP. It has therefore been suggested that GAP may provide a connection or link between growth factor receptors and the ras p21 family.Specifications
RASA1 | |
Monoclonal | |
Unconjugated | |
RASA1 | |
CM-AVM, CMAVM, GAP, PKWS, RASA, RASGAP, p120, p120GAP, p120RASGAP | |
Mouse | |
Affinity Chromatography | |
RUO | |
5921 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1H2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P20936 | |
RASA1 | |
Full length human recombit protein of human RASA1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.