Learn More
RASA1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305647AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat and Bovine
GTPase Activating Protein (GAP) is capable of stimulating the hydrolysis of GTP by ras p21 proteins, though GAP has little effect on the oncogenic forms of ras. It is also known that several tyrosine kinases such as platelet derived growth factor receptor and epidermal growth factor receptor are involved in the tyrosine phosphorylation of GAP. It has therefore been suggested that GAP may provide a connection or link between growth factor receptors and the ras p21 family.Specifications
RASA1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P20936 | |
RASA1 | |
Between 997 and 1047 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RASA1 | |
GAP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5921 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.