Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RASA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550740
Description
The antibody detects endogenous levels of total RASA2 protein.
The Ras-related low molecular mass GTPases participate in signal transduction involving a variety of cellular functions, including cell-cycle progression, cellular differentiation, cytoskeletal organization, protein transport and secretion. The Ras-GTPase-activating protein (RASGAP) forms a complex with a second protein, p190, in growth factor stimulated and tyrosinekinase transformed cells. The RASGAP p190 complex may serve to couple Ras- and Rho-mediated signaling pathways. RASGAP forms an abundant SH2-mediated complex with p190 RhoGAP in cells expressing activated tyrosine kinases.
Specifications
RASA2 | |
Polyclonal | |
Unconjugated | |
RASA2 | |
5430433H21Rik; AA517451; AU023900; CMAVM; CM-AVM; DKFZp434N071; GAP; GAP1M; GTPase-activating protein 1m; GTPase-activating protein of RAS; p120GAP; p120RASGAP; PKWS; ras GTPase-activating protein 2; RAS p21 protein activator 2; RASA; Rasa2; RASGAP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5922 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
3 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q15283 | |
RASA2 | |
Synthetic peptide corresponding to a region derived from internal residues of human RAS p21 protein activator 2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction