Learn More
Invitrogen™ RASA3 Polyclonal Antibody

Description
Recommended positive controls: A431, H1299, HeLa, HepG2. Predicted reactivity: Chicken (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | RASA3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | RASA3 |
| Gene Accession No. | Q14644 |
| Gene Alias | AI326412; C86362; E130011G04; GAP1(IP4BP); GAP1-InsP4 BP; GAP1IP4BP; GAPIII; GAPIII activator 3; GTPase activating protein III; GTPase-activating protein 1 family, inositol 1,3,4,5-tetrakisphosphate-binding protein; ins P4-binding protein; Ins(1,3,4,5)P4-binding protein; ras GTPase-activating protein 3; Ras GTPase-activating protein III; RAS p21 protein activator 3; Rasa3; RP11-245B11.3; R-Ras gap; R-Ras GTP activating protein; R-ras GTPase activating protein; R-ras RTPase activating protein-3; scat |
| Gene Symbols | RASA3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.