Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RASA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531934
Description
Recommended positive controls: HepG2. Predicted reactivity: Mouse (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the GAP1 family of GTPase-activating proteins that suppresses the Ras/mitogen-activated protein kinase pathway in response to Ca(2+). Stimuli that increase intracellular Ca(2+) levels result in the translocation of this protein to the plasma membrane, where it activates Ras GTPase activity. Consequently, Ras is converted from the active GTP-bound state to the inactive GDP-bound state and no longer activates downstream pathways that regulate gene expression, cell growth, and differentiation. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
RASA4 | |
Polyclonal | |
Unconjugated | |
RASA4 | |
AA793972; AW112107; BB079060; Ca2+promoted Ras inactivator; Ca2+-promoted Ras inactivator; calcium-promoted RAS inactivator; Capri; GAPL; Kiaa0538; Ras GTPase-activating protein 4; RAS p21 protein activator 4; RASA4; RasGAP-activating-like protein 2; RGD1565457 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10156 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.71 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O43374 | |
RASA4 | |
Recombinant protein encompassing a sequence within the N-terminus region of human RASA4. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction