Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560197
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560197 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560197 Supplier Invitrogen™ Supplier No. PA560197
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSSSFKKLYF SLTTEALSFA KTPSSKKSAL IKLANIRAAE KVEEKSFGGS HVMQVIYTDD AGRPQTAYLQ CKC Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.

This gene encodes a member of the GAP1 family of GTPase-activating proteins that suppresses the Ras/mitogen-activated protein kinase pathway in response to Ca(2+). Stimuli that increase intracellular Ca(2+) levels result in the translocation of this protein to the plasma membrane, where it activates Ras GTPase activity. Consequently, Ras is converted from the active GTP-bound state to the inactive GDP-bound state and no longer activates downstream pathways that regulate gene expression, cell growth, and differentiation. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASA4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RASA4
Gene Accession No. O43374
Gene Alias AA793972; AW112107; BB079060; Ca2+promoted Ras inactivator; Ca2+-promoted Ras inactivator; calcium-promoted RAS inactivator; Capri; GAPL; Kiaa0538; Ras GTPase-activating protein 4; RAS p21 protein activator 4; RASA4; RasGAP-activating-like protein 2; RGD1565457
Gene Symbols RASA4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RASA4. Recombinant protein control fragment (Product #RP-99250).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10156
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.