Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110274
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110274 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110274 Supplier Invitrogen™ Supplier No. PA5110274
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HLPPTFHAVA FYVMDEDALS RDDVIGKVCL TRDTIASHPK GFSGWAHLTE VDPDEEVQGE IHLRLEVWPG ARACRLRCSV LEARDLAPKD RNGTSDPFVR VRYKGRTRET SIVKKSCYPR WNETFEFELQ EGAMEALCVE AWDWDLVSRN DFLGKVVIDV QRLRVVQQEE GWFRLQPDQS KSRRHDEGNL GSLQLEVRLR DETVLPSSYY QPLVHLLCHE VKLGMQGPGQ L.

This gene encodes a member of the GAP1 family of GTPase-activating proteins that suppresses the Ras/mitogen-activated protein kinase pathway in response to Ca(2+). Stimuli that increase intracellular Ca(2+) levels result in the translocation of this protein to the plasma membrane, where it activates Ras GTPase activity. Consequently, Ras is converted from the active GTP-bound state to the inactive GDP-bound state and no longer activates downstream pathways that regulate gene expression, cell growth, and differentiation. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASA4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RASA4
Gene Accession No. O43374, Q6PFQ7
Gene Alias AA793972; AW112107; BB079060; Ca2+promoted Ras inactivator; Ca2+-promoted Ras inactivator; calcium-promoted RAS inactivator; Capri; GAPL; Kiaa0538; Ras GTPase-activating protein 4; RAS p21 protein activator 4; RASA4; RasGAP-activating-like protein 2; RGD1565457
Gene Symbols RASA4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 60-290 of human RASA4 (NP_008920.5).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10156, 288589, 54153
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.