Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASA4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5115826

Catalog No. PIPA5115826


Only null left
Add to Cart

Description

Description

Antibody detects endogenous levels of total RASA4.

This gene encodes a member of the GAP1 family of GTPase-activating proteins that suppresses the Ras/mitogen-activated protein kinase pathway in response to Ca(2+). Stimuli that increase intracellular Ca(2+) levels result in the translocation of this protein to the plasma membrane, where it activates Ras GTPase activity. Consequently, Ras is converted from the active GTP-bound state to the inactive GDP-bound state and no longer activates downstream pathways that regulate gene expression, cell growth, and differentiation. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RASA4
Polyclonal
Unconjugated
RASA4
AA793972; AW112107; BB079060; Ca2+promoted Ras inactivator; Ca2+-promoted Ras inactivator; calcium-promoted RAS inactivator; Capri; GAPL; Kiaa0538; Ras GTPase-activating protein 4; RAS p21 protein activator 4; RASA4; RasGAP-activating-like protein 2; RGD1565457
Rabbit
Affinity Chromatography
RUO
10156, 54153
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide
O43374, Q6PFQ7
RASA4
A synthesized peptide derived from human RASA4(Accession O43374), corresponding to amino acid residues V671-L721.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.