Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASAL1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550867

Catalog No. PA550867


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total RASAL1 protein.

The protein encoded by this gene is member of the GAP1 family of GTPase-activating proteins. These proteins stimulate the GTPase activity of normal RAS p21 but not its oncogenic counterpart. Acting as a suppressor of RAS function, the protein enhances the weak intrinsic GTPase activity of RAS proteins resulting in the inactive GDP-bound form of RAS, thereby allowing control of cellular proliferation and differentiation. This particular family member contains domains which are characteristic of the GAP1 subfamily of RasGAP proteins but, in contrast to the other GAP1 family members, this protein is strongly and selectively expressed in endocrine tissues. Alternatively spliced transcript variants that encode different isoforms have been described.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RASAL1
Polyclonal
Unconjugated
Rasal1
GAP1 like protein; MRASAL; Ras GTPase-activating-like protein; RAS protein activator like 1; RAS protein activator like 1 (GAP1 like); Rasal; Rasal1; RasGAP-activating-like protein 1
Rabbit
Antigen affinity chromatography
RUO
8437
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1.2 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
O95294
Rasal1
Fusion protein corresponding to residues near the N terminal of human RAS protein activator like 1 (GAP1 like).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.