Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASAL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559664
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559664 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559664 Supplier Invitrogen™ Supplier No. PA559664
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RTHSAVTLGD WSDPLDPDAE AQTVYRQLLL GRDQLRLKLL EDSNMDTTLE ADTGACPEVL ARQRAATARL LEVLADLDRA HEEFQQQER Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 70%.

The protein encoded by this gene is member of the GAP1 family of GTPase-activating proteins. These proteins stimulate the GTPase activity of normal RAS p21 but not its oncogenic counterpart. Acting as a suppressor of RAS function, the protein enhances the weak intrinsic GTPase activity of RAS proteins resulting in the inactive GDP-bound form of RAS, thereby allowing control of cellular proliferation and differentiation. This particular family member contains domains which are characteristic of the GAP1 subfamily of RasGAP proteins but, in contrast to the other GAP1 family members, this protein is strongly and selectively expressed in endocrine tissues. Alternatively spliced transcript variants that encode different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASAL1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rasal1
Gene Accession No. O95294
Gene Alias GAP1 like protein; MRASAL; Ras GTPase-activating-like protein; RAS protein activator like 1; RAS protein activator like 1 (GAP1 like); Rasal; Rasal1; RasGAP-activating-like protein 1
Gene Symbols Rasal1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RASAL1. Recombinant protein control fragment (Product #RP-97697).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8437
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.