Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RASAL2 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP119150JF549
Description
RASAL2 Polyclonal antibody specifically detects RASAL2 in Human samples. It is validated for Western BlotSpecifications
RASAL2 | |
Polyclonal | |
Western Blot | |
MGC129919, NGAP, ras GTPase-activating protein nGAP, Ras protein activator like 1, RAS protein activator like 2, RAS protein activator-like 2 | |
The immunogen recognized by this antibody maps to a region between residue 888 and 938 of human RAS protein activator like 2 using the numbering given in entry NP_004832.1 (GeneID 9462). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
9462 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction