Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASAL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590235
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590235 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590235 Supplier Invitrogen™ Supplier No. PA590235
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LADITKSLTN PTPIQQQLRR FTEHNSSPNV SGSLSSGLQK IFEDPTDSDL HKLKSPSQDN TDSYFRGKTL LLVQQASSQS MTYSEKDERE SSLPNGRSVS LMDLQDTHAA QVEHASVMLD VPIRLTGSQL SITQVASIKQ LRETQSTPQS APQVRRPLHP ALNQPGGLQP LSFQNPVYHL NNPIPAMPKA SIDSSLENLS TASSRSQSNS EDFKLSGPSN SSMEDFTKRS TQSEDFSRRH TVPDRHIPLA LPRQNSTGQA QIRKVDQGGL GARAKAPPSL PHSASLRSTG S; Positive Samples: HeLa.

This gene encodes a protein that contains the GAP-related domain (GRD), a characteristic domain of GTPase-activating proteins (GAPs). GAPs function as activators of Ras superfamily of small GTPases. The protein encoded by this gene is able to complement the defective RasGAP function in a yeast system. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASAL2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RASAL2
Gene Accession No. Q9UJF2
Gene Alias 6530401P13; A330066M24Rik; NGAP; Ras GTPase-activating protein nGAP; Ras protein activator like 1; RAS protein activator like 2; RAS protein activator-like 2; RASAL2
Gene Symbols RASAL2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 750-1040 of human RASAL2 (NP_7337932).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 304893, 9462
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.