Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RASD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571781
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571781 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571781 Supplier Invitrogen™ Supplier No. PA571781
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RASD1 Polyclonal Antibody for Western Blot, IHC (P)

Small GTPase. Negatively regulates the transcription regulation activity of the APBB1/FE65-APP complex via its interaction with APBB1/FE65.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RASD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RASD1
Gene Accession No. O35626, Q9Y272
Gene Alias Activator of G-protein signaling 1; AGS1; Dexamethasone-induced Ras-related protein 1; DEXRAS1; DEXRAS1 (Dexras1); MGC:26290; ras related dexamethasone induced 1; RAS, dexamethasone-induced 1; Rasd1; ras-related protein
Gene Symbols RASD1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 248-277 amino acids from the C-terminal region of human RASD1.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19416, 51655
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.