Learn More
Invitrogen™ RASD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120364
Description
Positive test controls include: A-549, LO2, 293T, Mouse brain, Mouse heart, Mouse kidney. The target is usually found in the following locations: Cell membrane, Cytoplasm, Cytoplasmic side, Lipid-anchor, Nucleus, perinuclear region. Immunogen sequence: TKENVDVPLV ICGNKGDRDF YREVDQREIE QLVGDDPQRC AYFEISAKKN SSLDQMFRAL FAMAKLPSEM SPDLHRKVSV QYCDVLHKKA LRNKKLLRAG SGGGGGDPGD AFGIVAPFAR RPSVHSDLMY IREKASAGSQ AKDKERCVIS.
Small GTPase. Negatively regulates the transcription regulation activity of the APBB1/FE65-APP complex via its interaction with APBB1/FE65.
Specifications
RASD1 | |
Polyclonal | |
Unconjugated | |
RASD1 | |
Activator of G-protein signaling 1; AGS1; Dexamethasone-induced Ras-related protein 1; DEXRAS1; DEXRAS1 (Dexras1); MGC:26290; ras related dexamethasone induced 1; RAS, dexamethasone-induced 1; Rasd1; ras-related protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
19416, 51655 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.78 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O35626, Q9Y272 | |
RASD1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 132-281 of human RASD1 (NP_057168.1). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.