Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RASEF Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501727274 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7274 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7274 Supplier Proteintech Group Inc Supplier No. 115691AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Binds predomitly GDP, and also GTP.

Specifications

Antigen RASEF
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RASEF
Gene Accession No. Q5RI75, Q8IZ41
Gene Alias RAB45, Ras related protein Rab 45, RASEF
Gene Symbols RASEF
Host Species Rabbit
Immunogen RASEF Fusion Protein Ag2149
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158158, 242505
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.