Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RASGEF1A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531598
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (91%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Guanine nucleotide exchange factor (GEF) with specificity for RAP2A, KRAS, HRAS, and NRAS (in vitro). Plays a role in cell migration.
Specifications
RASGEF1A | |
Polyclonal | |
Unconjugated | |
RASGEF1A | |
6330404M18Rik; AI835194; CG4853; CG4853 gene product; RasGEF domain family member 1A; RasGEF domain family, member 1A; ras-GEF domain-containing family member 1A; Rasgef1a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
221002 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8N9B8 | |
RASGEF1A | |
Recombinant fragment corresponding to a region within amino acids 20 and 302 of Human RASGEF1A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction