Learn More
Invitrogen™ RASGEF1A Polyclonal Antibody

Description
The antibody detects endogenous levels of total RASGEF1A protein.
Specifications
Specifications
| Antigen | RASGEF1A |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.7 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | RASGEF1A |
| Gene Accession No. | Q8N9B8 |
| Gene Alias | 6330404M18Rik; AI835194; CG4853; CG4853 gene product; RasGEF domain family member 1A; RasGEF domain family, member 1A; ras-GEF domain-containing family member 1A; Rasgef1a |
| Gene Symbols | RASGEF1A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.